Culti-Loops™ Shigella sonnei Gp D ATCC™ 25931™
Leverandør: THERMO FISHER DIAGNOSTICS
R4608150
OXOIR4608150EA 3225 NOK
OXOIR4608150
Culti-Loops™ Shigella sonnei Gp D ATCC™ 25931™
Mikrobiologiske testsystemer Tester for mikroorganismer
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium.
Forpakning: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.