Culti-Loops™ Salmonella enterica subsp. enterica serovar Enteritidis ATCC™ 13076™
Leverandør: THERMO FISHER DIAGNOSTICS
R4608200
OXOIR4608200EA 3225 NOK
OXOIR4608200
Culti-Loops™ Salmonella enterica subsp. enterica serovar Enteritidis ATCC™ 13076™
Mikrobiologiske testsystemer Tester for mikroorganismer
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Forpakning: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.