Culti-Loops™ Neisseria gonorrhoeae ATCC™ 49226™
Leverandør: THERMO FISHER DIAGNOSTICS
R4609006
OXOIR4609006EA 3225 NOK
OXOIR4609006
Culti-Loops™ Neisseria gonorrhoeae ATCC™ 49226™
Mikrobiologiske testsystemer Tester for mikroorganismer
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Forpakning: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.