Lactic acid 10% vandig løsning

Leverandør: THERMO FISHER DIAGNOSTICS
Danger

SR0021H SR0021K
OXOISR0021HEA 620 NOK
OXOISR0021H OXOISR0021K
Lactic acid 10% vandig løsning
Lactic acid
Formel: H₃CCH(OH)COOH
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004520
CAS-nummer: 50-21-5
EINECS: 200-018-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR