Penicillin G sodium salt

Leverandør: Biowest

Synonymer: 6-(2-Phenylacetamido)penicillanic acid sodium salt, Benzylpenicillin sodium salt, Sodium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate

P0018-1MU
P0018-1MUEA 160 NOK
P0018-1MU
Penicillin G sodium salt
Penicillin G sodium salt

Penicillin G sodium salt inhibits the final stages of bacteria cell wall synthesis. It inhibits primarily Gram-positive bacteria.


  • Storage temperature of 2 to 8 °C
  • pH of 5,5 to 7,5
  • Shelf life of 36 months


NB: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.

Formel: C₁₆H₁₇N₂NaO₄S
MDL nummer: MFCD00069666
CAS-nummer: 69-57-8

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR