Zinkoksid Ph. Eur., Macron Fine Chemicals™

Leverandør: Avantor Performance Materials

P106-33EA 540 NOK
P106-33 P106-42
Zinkoksid Ph. Eur., Macron Fine Chemicals™
Zinc oxide

Sertifiseringer: Meets Ph. Eur. Chemical Specifications.

Formel: ZnO
MDL nummer: MFCD00011300
CAS-nummer: 1314-13-2
FN: 3077
ADR: 9,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR