Zinksulfat heptahydrat Ph. Eur., Macron Fine Chemicals™

Leverandør: Avantor Performance Materials

P134-33EA 1125 NOK
P134-33 P134-38
Zinksulfat heptahydrat Ph. Eur., Macron Fine Chemicals™
Zinksulfat heptahydrat

Sertifiseringer: Meets Ph. Eur. chemical specifications.

Formel: ZnSO₄·7H₂O
MDL nummer: MFCD00149894
CAS-nummer: 7446-20-0
FN: 3077
ADR: 9,III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR