tri-Natriumcitrat dihydrat Ph. Eur., Macron Fine Chemicals™

Leverandør: Avantor Performance Materials

P149-33EA 770 NOK
P149-33 P149-38
tri-Natriumcitrat dihydrat Ph. Eur., Macron Fine Chemicals™
tri-Natriumcitrat dihydrat
Formel: Na₃C₆H₅O₇·2H₂O
MDL nummer: MFCD00150031
CAS-nummer: 6132-04-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR