Gentamicin sulphate

Leverandør: Biowest

Synonymer: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

P4020-1GR P4020-5GR
P4020-1GREA 565 NOK
P4020-1GR P4020-5GR
Gentamicin sulphate
Gentamicin sulphate

Gentamycin sulphate acts against Gram-negative, Gram-positive bacteria and mycoplasma.


  • Storage at 2 to 8 °C
  • Shelf life of 48 months


Used in cell culture applications at 1 ml/l. This concentration is for tissue culture media containing serum; serum-free media generally require lower concentration.


NB: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.

MDL nummer: MFCD00270181
CAS-nummer: 1405-41-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR