Tool cart
Leverandør: Palbam
CT0702
PALBCT0702EA 70775 NOK
PALBCT0702
Tool cart
Traller
Tool cart in stainless steel 304 with electropolished finish.
  • Five fully extendable drawers
  • One lock which secures all the drawers
  • Top ledge to prevent tools/parts falling
  • Cleanroom castors with brakes