Particle counter transport cart
Leverandør: Palbam
SENSORCART SENSORCARTARM PCCS0701 PCCS0902 PCCS0702 PCCS0722
139-0027EA 0 NOK
139-0027 139-0028 139-0587 PALBPCCS0902 PALBPCCS0702 PALBPCCS0722
Particle counter transport cart
Traller
This cart is specially designed for transporting particle sensing machines around the cleanroom environment. Stainless steel 304 with electropolished finish.
  • Ergonomic design
  • Shock absorbing system in each wheel protects the valuable particle sensor from vibrations and shocks