Wafer casettes
Leverandør: Palbam
PCPC0101
PALBPCPC0101EA 24450 NOK
PALBPCPC0101
Wafer casettes
Wafer håndtering Wafer kassetter
The palbam class stainless steel wafer cassette is designed to replace standard plastic cassettes, in the semiconductor manufacturing process. The cassette has been designed with H Bar.
  • Non-contaminating
  • Non-deforming
  • Fully conductive
  • Withstands temperatures up to 400 ºC
  • Designed for both manual and robotic handling