OmniTop Sample Tubes® Adjustable Volume Sampling System (AVSS)
Leverandør: Avantor Fluid Handling
PAWBAVSS15PP00EA 99275 NOK
PAWBAVSS15PP00 PAWBAVSS15PP00-KIT PAWBAVSS15PP01 PAWBAVSS15PP01-KIT
OmniTop Sample Tubes® Adjustable Volume Sampling System (AVSS)
Rør Reagensrør
Enhance and optimise sampling processes with the novel Adjustable Volume Sampling Systems (AVSS) from Avantor featuring OmniTop Sample Tubes®.
  • Customised sample volume at point of use
  • Aseptic, closed system sampling reduces contamination risk
  • Improved product yield
  • Reduced assembly and installation time
  • Easy operation and transfer
  • Single-use assemblies are available with a C-Flex® or PVC (Tygon®) tubing inlet option
The Adjustable Volume Sampling System (AVSS) can be used as a stand-alone sampling and transfer device or is custom configurable intoMore Product Information