Avantor SterilEnz®-II/EC: pre-gasketed polypropylene end caps
Leverandør: Avantor Fluid Handling
PAWBGTZ-EC-150-10EA 2900 NOK
PAWBGTZ-EC-150-10 PAWBSTZ-EC-150-01 PAWBSTZ-EC-150-10
Avantor SterilEnz®-II/EC: pre-gasketed polypropylene end caps
Fittings og kontakter
SterilEnz®-II/EC PP end caps are the only end caps that come equipped with a medical grade silicone gasket pre-attached to the fitting face. This feature prevents operators from misaligning or dropping gaskets during assembly. SterilEnz®-II/EC caps work with any standard flange gasket as well. Simply remove the pre-attached gasket (no tools required) and install your own.
  • Standard sanitary clamp fitting per ASME BPE specifications
  • Certified free from any animal derived components or processes
  • Gamma-stable, for use in irradiated disposable systems, 50 kGy