Human acute myeloid leukemia (AML) plasma
Leverandør: Charles River Laboratories Cell Solutions
PB006C-1-AML
PB006C-1-AMLEA 0 NOK
PB006C-1-AML
Human acute myeloid leukemia (AML) plasma
Plasma
Plasma used for researching the causes, diagnosis, supportive care and treatment of acute myeloid leukemia (AML). Collected from IRB-consented volunteer donors blood diagnosed with acute myeloid leukemia (AML).
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Store at −80 °C.
NB: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.