Nitrogen Generator, Horizen 24
Leverandør: PEAK SCIENTIFIC
3304532
PEAK3304532EA 0 NOK
PEAK3304532
Nitrogen Generator, Horizen 24
Gassgeneratorer Nitrogengeneratorer
Bringing Single Quad LC-MS users the very latest in nitrogen gas generator technology, Horizen 24 is the culmination of over 25 years at the forefront of nitrogen gas generation for the laboratory.
  • The most energy-efficient nitrogen generator on the market
  • Ultra-dry, non-methane hydrocarbon-free nitrogen at up to 99,9% purity and up to 116 psi
  • 55% less energy, saving power consumption*
  • Reduction in heat output by over 50%, saving on air conditioning costs*
  • Two year comprehensive manufacturer's warranty**
  • The smallest nitrogen generator in its class,
More Product Information
New Product