Air dryer, AD140L
Leverandør: PEAK SCIENTIFIC
40-1142EU 40-1142UK
PEAK40-1142EUEA 43650 NOK
PEAK40-1142EU PEAK40-1142UK
Air dryer, AD140L
Filter Gassfilter
The AD140L is a useful attachment for any laboratory's external source of air, removing moisture and ensuring the delivery of dry air. Moisture content in house air can contaminate lab instrumentation, impacting on results and in some cases damaging the instrument itself. Compact in size, wall-mountable and light, the AD140L requires little space and its few moving parts mean minimal maintenance is required.
  • Produces clean, dry air, ensures optimum supply for gas generators and instruments
  • Dryer performs 24/7 if needed
  • Dryer is wall-mountable for small