Air dryer, AD302L
Leverandør: PEAK SCIENTIFIC
40-1220EU 40-1220UK
PEAK40-1220EUEA 53725 NOK
PEAK40-1220EU PEAK40-1220UK
Air dryer, AD302L
Filter Gassfilter
The AD302L is a useful attachment for any laboratory's external source of air, removing moisture and ensuring the delivery of dry air. Moisture content in house air can contaminate lab instrumentation, impacting on results and in some cases damaging the instrument itself. Compact in size, wall-mountable and light, the AD302L requires little space and its few moving parts mean minimal maintenance is required.
  • Produces clean, dry air, ensures optimum supply for gas generators and instruments
  • Dryer performs 24/7 if needed
  • Dryer is wall-mountable for small
More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Fitting size 1×1/4" BSP Female
Vekt 20,5 kg
Elektronisk 240 VAC, 50/60 Hz
Trykk 6,89 - 8,27 bar (Inlet)
6,55 - 7,93 bar (Outlet)