Zero air generators, Precision Zero Air 1.5 L
Leverandør: PEAK SCIENTIFIC
60-1500
PEAK60-1500EA 36400 NOK
PEAK60-1500
Zero air generators, Precision Zero Air 1.5 L
Gassgeneratorer Luftfri generator
The Precision Zero Air generators are designed specifically to supply clean, dry, hydrocarbon-free air to be used as flame support gas for GC at both standard and trace detection limits.
  • Integrates seamlessly with other Precision units
  • Hydrocarbon content <0,05 ppm for market-leading purity
  • Minimum lifetime maintenance requirements, no expensive catalyst chamber replacements
  • Avoid risk of contaminants entering the system when switching out empty cylinders
  • No risk of running out of gas in middle of analysis
  • Compatible with Precision Air Compressor
  • Status