Nitrogen generators, Corona Nitrogen 1010
Leverandør: PEAK SCIENTIFIC
81-4000
PEAK81-4000EA 65425 NOK
PEAK81-4000
Nitrogen generators, Corona Nitrogen 1010
Gassgeneratorer Nitrogengeneratorer
The Corona 1010 meet the specifications of the Thermo Scientific Corona Veo Charged Aerosol Detector. This system can be purchased as a single nitrogen unit for laboratories with an external air supply or with a stackable air compressor for laboratories without an external air supply.
  • Suitable for applications requiring high purity nitrogen
  • Available with or without an external compressor
  • Tried and tested membrane technology delivers laboratory-grade nitrogen
  • Gas is supplied on demand so generator works to your schedule
  • 1 year comprehensive on-site