Hydraulic pellet press, KBr
Leverandør: PerkinElmer
51004599
PERC51004599EA 155000 NOK
PERC51004599
Hydraulic pellet press, KBr
Presser
This compact hydraulic press is used with the KBr die to prepare KBr pellets.
Pressure is produced by operating a pump lever.
Bestillingsinformasjon: Evacuable KBr die, KBr pellet holder and adjustable adapter have to be ordered separately.