Polystyrene Test Film - MIR
Leverandør: PerkinElmer
L1202057
PERCL1202057EA 770 NOK
PERCL1202057
Polystyrene Test Film - MIR
Standarder kromatografi standarder
This test film is an easy-to-use standard to check frequency, accuracy and any changes.
This reference material mounts easily in a FT-IR spectrometer, allowing the user to quickly and easily check the mid infrared frequencies and overall accuracy of the FT-IR spectrometer.
The Polystyrene Test Film is 0.038 mm thickness and untraceable.