Deuterium lamp, TruLife™ 1,5
Leverandør: PerkinElmer
L6022728
PERCL6022728EA 25125 NOK
PERCL6022728
Deuterium lamp, TruLife™ 1,5
Lys Deuterium lampe
These deuterium lamps provide the best wavelength coverage across the working range of the spectrometer for consistently high performance.
  • Lamps are pre-aligned for easy installation
  • Extended lifetime of 1500 hours
  • Built-in digital timer to monitor lamp lifetime