Tin capsule kit, large
Leverandør: PerkinElmer
N2411362
PERCN2411362EA 5125 NOK
PERCN2411362
Tin capsule kit, large
Prøvecontainere Beholdere, generelt bruk
Large tin capsule used for the analysis of nonvolatile solid samples. It is mostly used in the combustion of the sample.
Tin capsules are routinely used to analyse polymers, soils, coal, dry dairy products, plant leaves, paper, textiles, calibration standards, and many organic and pharmaceutical products.