Silver capsule for oxygen analysis kit
Leverandør: PerkinElmer
N2411364
PERCN2411364EA 8375 NOK
PERCN2411364
Silver capsule for oxygen analysis kit
Prøvecontainere Beholdere, generelt bruk
Silver capsule used for analysing organic substances. For oxygen content either silver or standard tin capsules may be used for analysing organic substances for oxygen content.