Hafnium 1.000 mg/l Hf in 2% saltsyre (fra HfOCl2) standard til AAS og ICP
Leverandør: PerkinElmer
N9300122
PERCN9300122EA 2925 NOK
PERCN9300122
Hafnium 1.000 mg/l Hf in 2% saltsyre (fra HfOCl2) standard til AAS og ICP
Standarder Kationstandarder Hafnium Standards
Pure Grade standard for AA and ICP-OES
  • Analysed by ICP-OES
  • Analysed by Classical Wet Assay
  • 32 trace impurities analyzed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppm level
  • All Standards are prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA