Calibration standard, single element, germanium for AAS and ICP-OES, pure grade
Leverandør: PerkinElmer
N9303774
PERCN9303774EA 1525 NOK
PERCN9303774
Calibration standard, single element, germanium for AAS and ICP-OES, pure grade
Standarder Kationstandarder Germanium Standards
NB: These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.