Watch glasses
Leverandør: PerkinElmer
N9308003
PERCN9308003EA 9800 NOK
PERCN9308003
Watch glasses
Urglass
Disposable watch glasses to promote gentle refluxing. For use with 50 ml and 100 ml DigiTUBEs.