AccuForge Gem® Scientific Blade Dispenser
Leverandør: PERSONNA MEDICAL
AGBL-7027-0000
PERSAGBL-7027-0000EA 31450 NOK
PERSAGBL-7027-0000
AccuForge Gem® Scientific Blade Dispenser
Blade Removers and Dispensers
AccuForge Pop-Up Dispenser provides quick, easy blade access for high volume applications. Each disposable dispenser contains 100 blades constructed of .009" carbon with aluminum back steel and 3-facet edges for sharpness.
  • Safety Dispenser with one piece disposable design
  • 3-facet blade edge for increased sharpness
  • Pop-up dispenser with easy push-button operation provides safe and convenient access to fresh blades
Wear eye protection. Handle and use with care. Keep out of children’s reach. Dispose of safely.
NB: Blades are extremely sharp. BendingMore Product Information