Natriumbenzoat, Supelco®

Leverandør: Merck

Synonymer: Benzoic acid sodium salt

PHR1231-1G
PHR1231-1GEA 910 NOK
PHR1231-1G
Natriumbenzoat, Supelco®
Natriumbenzoat
Formel: C₆H₅COONa
MW: 144,11 g/mol
Smeltepunkt: 410…430 °C
Tetthet: 1,44 g/cm³ (20 °C)
Flash punkt: >100 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012463
CAS-nummer: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Indeks: 13,08654
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=162cd8e62172f766c472321c2c2f0b7fa5361379&vid=162cd8e62172f766c472321c2c2f0b7fa5361379&prid=web&clid=SAPDAM

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR