Pharmaceutical secondary standard, Monobasic Potassium Phosphate, certified reference material, Supelco®
Leverandør: Merck
PHR1330-5G
PHR1330-5GEA 1575 NOK
PHR1330-5G
Pharmaceutical secondary standard, Monobasic Potassium Phosphate, certified reference material, Supelco®
Standarder Organiske standarder
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Sertifiseringer: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cc8f6fd2651458fd844ae74541fafb34d0a4092&vid=3cc8f6fd2651458fd844ae74541fafb34d0a4092&prid=web&clid=SAPDAM