Supelco® Organic Reference Standard, Ibuprofen Related Compound J

Leverandør: Merck

PHR1978-50MG
PHR1978-50MGEA 7350 NOK
PHR1978-50MG
Supelco® Organic Reference Standard, Ibuprofen Related Compound J
Standarder Organiske standarder

  • GHS signal: GHS07


NB: P264 - P270 - P301 + P312 - P501

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9bddfa8ebcce2aa96f1104e976144cbfe92066f&vid=a9bddfa8ebcce2aa96f1104e976144cbfe92066f&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) (2RS)-2-(4-Isobutyrylphenyl)propionic acid; (2-(4-Isobutyrylphenyl)propanoic acid); α-Methyl-4-(2-methyl-1-oxopropyl)benzeneacetic acid; (2RS)-2-[4-(2-Methylpropanoyl)phenyl]propanoic acid; 1-Oxoibuprofen
Environmentally Preferable
M-Clarity quality level MQ300
CAS-nr 65813-55-0
MDL No. MFCD08275584
Molekylvekt 220,26
Hardhetsklasse beskrivelse H302
Fraktforhold STD
Lagringsforhold 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR