L(-)-Ephedrine sulphate, certified reference material for pharmaceutical production

Leverandør: Merck
Warning

PHR8829-200MG
PHR8829-200MGEA 0 NOK
PHR8829-200MG
L(-)-Ephedrine sulphate, certified reference material for pharmaceutical production
L(-)-Ephedrine sulphate

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Formel: C₂₀H₃₂N₂O₆S
Lagringstemperatur: Fryseskap
CAS-nummer: 134-72-5
New Product

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR