Immunoassay microplates, Protein L coated plates, Pierce™
Leverandør: Thermo Fisher Scientific
15190
PIER15190EA 3700 NOK
PIER15190
Immunoassay microplates, Protein L coated plates, Pierce™
Mikroplater Immunologi-mikroplater og strips
Pierce™ Protein L coated plates provide an alternative to direct, passive adsorption methods for immobilising antibodies for ELISA and other plate-based assay techniques.
  • Retain antibody activity, which can be lost when antibodies are immobilised by passive adsorption
  • Orient antibodies for maximum antigen-binding capacity
  • Immobilise antibodies for plate assays without prior purification
  • Ensure minimal variation (<5% well-to-well) from consistent coating
  • Plates are pre-blocked with SuperBlock™ blocking buffer to reduce non specific binding
  • Bind 2 to 5