Chromatography cartridges, glutathione, Pierce™
Leverandør: Thermo Fisher Scientific
16109 16110
PIER16109EA 4325 NOK
PIER16109 PIER16110
Chromatography cartridges, glutathione, Pierce™
Kromatografikolonner
Pierce™ Glutathione chromatography cartridges are convenient, ready to use pre-packed devices for the purification of GST-fusion proteins from cellular lysates. The glutathione is immobilised through its central sulfhydryl onto 6% crosslinked agarose resin. Purification of GST-fusion proteins using glutathione (GSH) agarose beads is well documented and provides a one-step, high purity affinity purification.
  • Binds at least 40 mg of purified recombinant GST protein per ml of resin
  • Consistently purifies at least 10 mg of GST-tagged protein per ml of resin