Firefly luciferase glow assay kit, Pierce™

Leverandør: Thermo Fisher Scientific

16176 16177
PIER16176EA 1600 NOK
PIER16176 PIER16177
Firefly luciferase glow assay kit, Pierce™
Assay Proteinanalyser

The Pierce™ Firefly Luciferase Glow Assay Kit provides a bright and stable bioluminescence signal (half-life greater than 2 hours) in the presence of firefly luciferase reporters for protein expression assays.


  • Highly sensitive detection of firefly luciferase activity
  • Increased signal stability
  • Contains a universal cell lysis buffer and optimised glow assay reagent


The bioluminescent signal produced by firefly luciferase results from the oxidation of D-Luciferin. The Firefly Luciferase Glow Kit produces an extended light signal correlating with the amount of firefly protein expressed, which is proportional to the activity of the promoter for firefly expression.


Leveringsinformasjon: Kit contains Firefly Glow assay buffer (5 or 50 ml), D-Luciferin (3 or 30 mg) and 2X Cell Lysis Buffer (6 or 60 ml).


NB: For research use only. Not for use in diagnostic procedures. It stored at —20 °C

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR