Biotinylated-protein interaction pull down kit, Pierce™
Leverandør: Thermo Fisher Scientific
21115
PIER21115EA 7350 NOK
PIER21115
Biotinylated-protein interaction pull down kit, Pierce™
Assay Proteinanalyser
Pierce™ biotinylated protein interaction pull-down kit contains the necessary components to capture and purify interactors of a biotin-labeled protein or ligand.
  • Provides a complete easy-to-use strategy for discoverying protein:protein interactions
  • Uses common laboratory equipment and reagents
  • Adaptable to single- or multiple-sample demands
  • Flexible pull-down format uses spin cups for easy and efficient manipulation of streptavidin agarose beads
Leveringsinformasjon: Kit includes streptavidin agarose resin, 1,5 ml settled gel; lysis buffer, 250 ml; BupH tris buffered saline, 1 pack (500 ml); wash buffer (acetate, pH 5), 100 ml; elution buffer (pH 2,8), 50 ml; spin columns accessory pack, 27 columns; collection tubes and caps, 200×2 ml tubes.