Streptavidin poly-HRP conjugate, Pierce™
Leverandør: Thermo Fisher Scientific
21140
PIER21140EA 3450 NOK
PIER21140
Streptavidin poly-HRP conjugate, Pierce™
Assay Proteinanalyser
Pierce™ Streptavidin Poly-HRP conjugate is biotin-binding protein conjugated with polymers of horseradish peroxidase, enabling signal amplification and detection of biotinylated antibodies for IHC and other methods.
  • Detect low-abundant targets (low picogram-femtogram range) with high signal-to-noise ratios
  • Compatible with chromogenic, fluorogenic and chemiluminescent substrates, also with ELISA, Western blotting and immunohistochemistry (IHC)
  • Can be directly substituted into immunoassays and other detection assays
  • Requires less conjugate per assay and standard HRP conjugates