Sulpho-NHS Biotin, EZ-Link™ No-Weigh™ Format

Leverandør: Thermo Fisher Scientific

Synonymer: Biotin 3-sulpho-N-hydroxysuccinimide ester sodium salt, Sulphosuccinimidyl biotin

21217 A39256
PIER21217EA 3625 NOK
PIER21217 PIERA39256
Sulpho-NHS Biotin, EZ-Link™ No-Weigh™ Format
Sulpho-NHS Biotin

Thermo Scientific EZ-Link Sulfo-NHS-Biotin is a short-chain, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.


  • Protein labeling—biotinylate antibodies to facilitate immobilization, purification or detection using streptavidin resins or probes
  • Cell surface labeling—biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
  • Amine-reactive—reacts with primary amines (-NH2), such as lysine side-chains or the amino-termini of polypeptides
  • Soluble—charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
  • Irreversible—forms permanent amide bonds; spacer arm cannot be cleaved
  • Very short—spacer arm (total length added to target) is 13.5 angstroms; it consists of the native biotin valeric acid group only.


Sulfo-NHS-Biotin is the shortest of three very similar EZ-Link Reagents that are water-soluble, non-cleavable, and enable simple and efficient biotinylation of antibodies, proteins and any other primary amine-containing macromolecules in solution. Specific labeling of cell surface proteins is another common application for these uniquely water-soluble and membrane impermeable reagents. Differing only in their spacer arm lengths, the three Sulfo-NHS-ester reagents offer the possibility of optimizing labeling and detection experiments where steric hindrance of biotin binding is an important factor. Sulfo-NHS-Biotin is offered in several package sizes and as complete protein labeling kits.

Formel: C₁₄H₁₈N₃NaO₈S₂
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00078535
CAS-nummer: 119616-38-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR