Sulpho-NHS-SS-Biotin, EZ-Link™, Pierce™

Leverandør: Thermo Fisher Scientific

Synonymer: Biotinamidohexanoic acid 3-sulpho-N-hydroxysuccinimide ester sodium salt, Sulphosuccinimidyl 2-(biotinamido)-ethyl-1-3’-dithiopropionate

21331 A39258
PIER21331EA 6225 NOK
PIER21331 PIERA39258
Sulpho-NHS-SS-Biotin, EZ-Link™, Pierce™
Sulpho-NHS-SS-Biotin

Thermo Scientific EZ-Link Sulfo-NHS-SS-Biotin is a water-soluble, NHS-ester biotinylation reagent whose spacer arm includes a cleavable disulfide bond for reversible labeling of proteins and cell surface primary amines.


  • Protein labeling – biotinylate antibodies to facilitate immobilization, purification or detection
  • Cell surface labeling – biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
  • Amine-reactive—reacts with primary amines (-NH2), such as lysine side-chains, or the amino-termini of polypeptides
  • Cleavable—disulfide bond in spacer arm allows the biotin label to be removed using reducing agents such as DTT; only a small sulfhydryl group remains attached to the molecule
  • Soluble – charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
  • Medium length—spacer arm (total length added to target) is 24.3 angstroms; it consists of the native biotin valeric acid group extended by a 7-atom chain


Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging prevents the loss of reagent reactivity and contamination over time by eliminating the repetitive opening and closing of the vial. The format enables use of a fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.


Sulfo-NHS-SS-Biotin is a thiol-cleavable amine-reactive biotinylation reagent that contains an extended spacer arm to reduce steric hindrances associated with avidin binding. This reagent is water-soluble, enabling biotinylation to be performed in the absence of organic solvents such as DMSO or DMF for applications that cannot tolerate solvents or are complicated by their inclusion. The compound is particularly useful for labeling and purifying cell surface proteins, because (a) its sulfonate group prevents it from permeating cell membranes and (b) its cleavable spacer arm enables initially biotinylated proteins to be released from streptavidin affinity columns.


NB: For Research Use Only. Not for use in diagnostic procedures.

Formel: C₁₉H₂₇O₉N₄S₄Na
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00078533
CAS-nummer: 202057-28-1

Bestill nå

Spesifikasjon Testresultater

Description EZ-Link Sulfo-NHS-SS-Biotin
Product Type Sulfo-NHS-SS-Biotin
Quantity 100 mg
Product Line EZ-Link™
Cell Permeability Cell-Impermeant
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, Sulfo-NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Biotin
Label Type Biotin & Analogs
Spacer Mid-Length, Cleavable


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR