SIAB (N-Succinimidyl (4-iodoacetyl)aminobenzoate), Pierce™

Leverandør: Thermo Fisher Scientific

Synonymer: SIAB

22329
PIER22329EA 4525 NOK
PIER22329
SIAB (N-Succinimidyl (4-iodoacetyl)aminobenzoate), Pierce™
SIAB (N-Succinimidyl (4-iodoacetyl)aminobenzoate)

Thermo Scientific Pierce SIAB is a mid-length (10.6 angstrom) crosslinker for amine-to-sulfhydryl conjugation via N-hydroxysuccinimide (NHS) ester and iodoacetyl reactive groups.


  • Reactive groups: NHS ester and iodoacetyl
  • Reactive toward: amino and sulfhydryl groups
  • Non-cleavable
  • Water-insoluble (dissolve first in DMF or DMSO); compare to Sulfo-SIAB
  • Used to prepare stable enzyme-IgG conjugates
  • Forms conjugates with liposomes


NB: For Research Use Only. Not for use in diagnostic procedures.

Formel: C₁₃H₉IN₂O₅
MW: 400,125 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00078537
CAS-nummer: 72252-96-1

Bestill nå

Spesifikasjon Testresultater

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Haloacetyl, NHS Ester
Spacer Mid-Length
Form Powder
Quantity 50 mg
Type Crosslinker
Solubility DMF, DMSO

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR