Pierce™ Protein G Plus Agarose
Leverandør: Thermo Fisher Scientific
22851 22852
PIER22851EA 5500 NOK
PIER22851 PIER22852
Pierce™ Protein G Plus Agarose
Proteinrense Protein Purification Resins
Pierce™ protein G plus agarose is a high capacity protein A beaded agarose resin for use in a variety of antibody affinity purification methods.
  • Ideal for polyclonal IgG purification from mouse, human, cow, goat, and sheep serum, including human IgG3 and mouse IgG1 isotypes
  • Agarose resin support is crosslinked 6% beaded agarose (CL-6B)
  • Protein G made by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
  • High capacity beads with a high load of immobilised protein G
Binding capacity greater than 20 mg human IgG/ml resin.