Compat-Able™ Protein assay preparation reagent set
Leverandør: Thermo Fisher Scientific
23215
PIER23215EA 1975 NOK
PIER23215
Compat-Able™ Protein assay preparation reagent set
Assay Proteinanalyser
The Compat-Able™ Protein assay preparation reagent set eliminates interfering substances from protein assay samples to make them compatible with BCA and Bradford assays.
  • Removes nearly any substance that interferes with colorimetric protein assays: salts, detergents, denaturants, reducing agents, and even IEF sample buffer
  • Prepare samples in less than 10 minutes with four simple steps
  • Reagent set is easier to use than homemade TCA or acetone precipitation reagents, thereby producing more consistent results
  • Microcentrifuge method is easily scaled to larger centrifuge tube volumes for special needs
  • Protocol integrates control and normalisation procedures
Leveringsinformasjon: Kit contains sufficient reagents to pretreat up to 500 samples. It comprises Compat-Able™ Protein assay preparation reagent 1 (250 ml) and Compat-Able™ Protein assay preparation reagent 2 (250 ml).