Pierce™ Coomassie Plus (Bradford) Protein Assay Kit
Leverandør: Thermo Fisher Scientific
23236
PIER23236EA 3475 NOK
PIER23236
Pierce™ Coomassie Plus (Bradford) Protein Assay Kit
Assay Proteinanalyser
The Pierce™ Coomassie Plus Protein assay is a quick and ready-to-use, reducing agent-compatible, improved Bradford assay to quickly measure (A595 nm) total protein concentration compared to a protein standard.
  • Colorimetric measure with a standard spectrophotometer or plate reader (595 nm)
  • Easy to use single reagent; no working reagent preparation required
  • Fast almost immediate colour development; add, mix and read results
  • Broad range detects protein concentration in the range 1 to 1500 µg/ml
  • Better improved linearity and response-uniformity compared to traditional Bradford formulations
  • Flexible microplate and cuvette protocols provided with the instructions and adaptable to several target working ranges
This modification of the well-known Bradford method greatly reduces the tendency of Coomassie reagents to give non linear response curves by a formulation that substantially improves linearity for a defined range of protein concentration. In addition, the Coomassie Plus Reagent results in significantly less protein-to-protein variation than is observed with other Bradford-type Coomassie formulations.
Leveringsinformasjon: Kit contains sufficient reagents for 630 tube assays or 3160 microplate assays. Pierce™ Coomassie Plus™ Protein Assay Reagent, 950 ml; Albumin Standard Ampoules, 2 mg/ml, 10×1 ml.