Pierce™ GlycoProtein Carbohydrate Estimation Kits

Leverandør: Thermo Fisher Scientific

23259 23260
PIER23259EA 4075 NOK
PIER23259 PIER23260
Pierce™ GlycoProtein Carbohydrate Estimation Kits
Assay Proteinanalyser

Pierce™ Glycoprotein Carbohydrate Estimation Kit enables the amount of protein glycosylation to be measured as a percentage of total purified protein mass.


  • Enables quick and easy identification of an unknown protein sample as a glycoprotein
  • Assess sugar and carbohydrate contamination in non-glycosylated protein samples
  • Complementary to electrophoresis, Western blotting and ELISA-based procedures often used to detect glycoprotein
  • Determines carbohydrate content in three easy steps: oxidise, react and read
  • Entire assay can be performed in less than 75 minutes


Kit includes six purified glycoprotein standards and the required assay reagents to effectively estimate the amount of oxidisable glycosylation (percent carbohydrate by weight) of purified protein samples. Sugar groups in the glycoprotein sample are first oxidised with sodium meta-periodate to produce detectable aldehyde groups. Then the sample is reacted with the colorimetric glycoprotein detection reagent. Absorbance at 550 nm of the resulting purple reaction product is measured with a spectrophotometer or plate reader. Finally, the carbohydrate content in the glycoprotein is calculated by comparison to results from the five glycoprotein standards that are included in the kit.


Applications:
• Determine if a purified polyclonal antibody or other protein is glycosylated before attempting to perform carbohydrate-based conjugation or immobilization chemistries with hydrazide reagents
• Characterize and classify purified protein fractions from an affinity procedure
• Assess sugar and carbohydrate contamination in non-glycosylated protein samples


Leveringsinformasjon: Kit contains negative control (Lysozyme and BSA) 2,5 mg each, positive control (Ovalbumin) 2,5 mg each, apo-transferrin 2,5 mg, fetuin 0,25 mg, α1-acid glycoprotein 0,25 mg, aodium meta-periodate 500 mg, glycoprotein detection reagent 500 mg, and glycoprotein assay buffer 250 ml.

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Applikasjon Internal standard Solution-based detection, absorbance

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR