GelCode™ Blue Stain Reagent

Leverandør: Thermo Fisher Scientific

24590
PIER24590EA 1625 NOK
PIER24590
GelCode™ Blue Stain Reagent
Farger Elektroforese Farging

GelCode blue stain reagent is a ready-to-use protein stain based on colloidal coomassie dye G-250 that provides nanogram-level detection in polyacrylamide gels and for in-blot detection.


  • No methanol-acetic acid destaining required
  • Fast, one-step, one-hour staining ready-to-use stain reagent; band development is visible directly in the staining tray; no background from overnight staining; flexible fixing and washing protocol; no gel shrinkage; stained gel can be dried
  • Water wash increases staining sensitivity optional step provides crystal-clear gel background and makes weakly stained bands more visible


This unique GelCode blue stain reagent stains only protein and allows bands to be viewed directly in the gel during the one hour gel staining process. Because the destaining step does not fix the protein, GelCode blue stain is compatible with mass spectrometry analysis and N-terminal sequence analysis.


Detection location: In-blot detection, in-gel detection
Detection method: Colorimetric
Label or dye: Coomassie
Target molecule: Protein


Store at 4 °C.


Bestillingsinformasjon: The 6xhis protein tag staining kit (PIER24575) and phosphoprotein staining kit (PIER24550) come with the appropriate reagent and control sets, which can be purchased separately.


NB: For research use only. Not for use in diagnostic procedures.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR