PageBlue™ protein staining solution
Leverandør: Thermo Fisher Scientific
24620
PIER24620EA 1750 NOK
PIER24620
PageBlue™ protein staining solution
Farger Elektroforese Farging
PageBlue Protein Staining Solution is a sensitive, economical formulation of colloidal coomassie G-250 dye for endpoint staining of proteins in polyacrylamide gels and on PVDF membranes without methanol or acetic acid.
  • Easy-to-use—simply soak the gel in the ready-to-use solution and observe stained bands without destaining
  • Staining time—25 to 40 minute protocol
  • Safe—does not contain methanol or acetic acid
  • Dynamic range—linear dynamic range of 5 to 500 ng
  • Economical—can be reused up to three times
The simple PageBlue Protein Staining Solution protocol is sensitive, efficient, and eliminates the concern of over-staining gels, even after overnight staining. This protein stain delivers a dynamic range of 5 to 500 ng, which is approximately ten times more sensitive than traditional coomassie R-250-based dyes.
Bestillingsinformasjon: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store product at room temperature. Product shipped at ambient temperature