Immunoprecipitation, magnetic ChIP kit, Pierce™
Leverandør: Thermo Fisher Scientific
26157
PIER26157EA 9175 NOK
PIER26157
Immunoprecipitation, magnetic ChIP kit, Pierce™
Proteinrense Protein Purification Kits
The Pierce™ Magnetic ChIP Kit provides a convenient method for efficient isolation of chromatin-bound DNA by immunoprecipitation for subsequent quantitation by PCR.
  • Purified DNA for quantitative PCR can be obtained in about 8 hours
  • Optimised micrococcal nuclease digestion and nuclear lysis
  • Obtain results with as few as 10000 cells
The blocked protein A/G magnetic beads provide high binding capacity, low non specific background, and work with many antibody species. These beads can be used manually with a magnetic stand or with automated platforms.
Leveringsinformasjon: The kit contains sufficient reagents to perform 30 chromatin immunoprecipitation (ChIP) assays with appropriate controls using an optimised protocol. The kit includes reagents for cell lysis, capture of protein-DNA complexes, reversal of crosslinking, and DNA isolation.