Sulphosuccinimidyl acetate (Sulpho-NHS-acetate), Pierce™

Leverandør: Thermo Fisher Scientific

Synonymer: Sulpho-NHS-acetate, 1-(Acetyloxy)-2,5-dioxo-3-pyrrolidinesulphonic acid

26777
PIER26777EA 2425 NOK
PIER26777
Sulphosuccinimidyl acetate (Sulpho-NHS-acetate), Pierce™
Sulphosuccinimidyl acetate (Sulpho-NHS-acetate)

Pierce™ Sulfo-NHS-Acetate is a small compound used for blocking primary amines (eg. lysine side chains). This compound forms stable, covalent amide bonds with amine groups to prevent specific interactions or conjugations.


  • Irreversibly block primary amines
  • Soluble in DMSO, DMF and water
  • Perform reactions in standard non-amine-containing buffers at pH 7 or greater


Sulpho-NHS-Acetate is typically used to prevent polymerisation when performing protein crosslinking reactions and when conjugating peptides to carrier proteins for immunogen production. Blocking amines on the peptide allows directed conjugation of carboxylic acids on the peptide to primary amines on the protein using EDC.


NB: Upon receipt store desiccated at -20° C. Product is shipped at ambient temperature.

Formel: C₄H₄NNaO₆S
MDL nummer: MFCD01861959
CAS-nummer: 152305-87-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR