Iodination tubes, Pierce™
Leverandør: Thermo Fisher Scientific
28601
PIER28601EA 2450 NOK
PIER28601
Iodination tubes, Pierce™
Rør Reagensrør
Pierce™ Iodination Tubes are 12×75 mm glass test tubes that are pre-coated at the bottom with Pierce™ Iodination Reagent, which activates iodine-125 for protein or peptide iodination procedures.
  • Eliminates the tedious reagent surface coating step
  • Consistent, flake-resistant coating of the iodination reagent
  • Generates more than sufficient oxidative capacity for a typical protocol
  • Allows iodinations to be carried out directly in the tube or in two-steps
The pre-coated iodination tubes provide for convenient and efficient iodine labeling without theMore Product Information